Product Details

SNP ID
rs719516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17777670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTGTGTTTTGGGAAGGGATGAA[C/G]GGAAGGGAAGAGCAATGTGGGGAAT
Phenotype
MIM: 607642
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAI1 PubMed Links

Gene Details

Gene
RAI1
Gene Name
retinoic acid induced 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030665.3 Intron NP_109590.3
XM_017024025.1 Intron XP_016879514.1
XM_017024026.1 Intron XP_016879515.1
XM_017024027.1 Intron XP_016879516.1
XM_017024028.1 Intron XP_016879517.1
Gene
SMCR5
Gene Name
Smith-Magenis syndrome chromosome region, candidate 5 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details