Product Details

SNP ID
rs168818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138358376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATAAAGGGGCCATTAAATAACTC[A/G]TCTAAAAAAGAATTAGGCACTCTAG
Phenotype
MIM: 608435
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRAS PubMed Links

Gene Details

Gene
MRAS
Gene Name
muscle RAS oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085049.2 Intron NP_001078518.1
NM_001252090.1 Intron NP_001239019.1
NM_001252091.1 Intron NP_001239020.1
NM_001252092.1 Intron NP_001239021.1
NM_001252093.1 Intron NP_001239022.1
NM_012219.4 Intron NP_036351.3
XM_005247228.1 Intron XP_005247285.1
XM_005247229.2 Intron XP_005247286.1
XM_017005887.1 Intron XP_016861376.1

View Full Product Details