Product Details

SNP ID
rs626339
Assay Type
Functionally tested
NCBI dbSNP Submissions
56
Location
Chr.1:111186703 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
AGTGCAGCGATCCCTTCACCTTTAG[C/T]TAAAGAATTGGACTGTGCTGCTCAA
Phenotype
MIM: 616751 MIM: 615111
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEPT1 PubMed Links

Gene Details

Gene
CEPT1
Gene Name
choline/ethanolamine phosphotransferase 1
There are no transcripts associated with this gene.

Gene
DENND2D
Gene Name
DENN domain containing 2D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271833.1 2562 UTR 3 NP_001258762.1
NM_024901.4 2562 UTR 3 NP_079177.2
XM_006710921.2 2562 Intron XP_006710984.2
XM_006710922.1 2562 Intron XP_006710985.1
XM_006710923.1 2562 Intron XP_006710986.1
XM_006710924.1 2562 Intron XP_006710987.1
XM_011542187.1 2562 Intron XP_011540489.1
XM_011542189.1 2562 Intron XP_011540491.1
XM_011542190.2 2562 Intron XP_011540492.1
XM_017002386.1 2562 Intron XP_016857875.1
XM_017002387.1 2562 Intron XP_016857876.1
XM_017002388.1 2562 Intron XP_016857877.1
XM_017002389.1 2562 Intron XP_016857878.1

View Full Product Details