Product Details
- SNP ID
-
rs209231
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:130213832 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGCTGACCACCAAAGGGAAAAAAA[C/T]CCCTGCAAGCCAGGTGCTGAAGCCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF280C
PubMed Links
Gene Details
- Gene
- ZNF280C
- Gene Name
- zinc finger protein 280C
View Full Product Details