Product Details

SNP ID
rs566561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115832512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGTGAATTAACAATACTACAC[A/C]ATATACATATTATACAGATGTGCAT
Phenotype
MIM: 601507 MIM: 609608
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AP3S1 PubMed Links

Gene Details

Gene
AP3S1
Gene Name
adaptor related protein complex 3 sigma 1 subunit
There are no transcripts associated with this gene.

Gene
ATG12
Gene Name
autophagy related 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277783.1 Intron NP_001264712.1
NM_004707.3 Intron NP_004698.3

View Full Product Details