Product Details

SNP ID
rs2281922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:91220617 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGACCCTTGGAGCATCTTCAAAGC[A/G]TCATCAGAATTCCACAGGATCATAG
Phenotype
MIM: 600529
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AUH PubMed Links

Gene Details

Gene
AUH
Gene Name
AU RNA binding protein/enoyl-CoA hydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306190.1 Intron NP_001293119.1
NM_001698.2 Intron NP_001689.1
XM_005252066.3 Intron XP_005252123.1
XM_005252067.4 Intron XP_005252124.1
XM_005252069.4 Intron XP_005252126.1
XM_005252072.2 Intron XP_005252129.1
XM_006717150.3 Intron XP_006717213.1
XM_011518800.2 Intron XP_011517102.1
XM_011518802.2 Intron XP_011517104.1
XM_011518803.2 Intron XP_011517105.1
XM_011518804.2 Intron XP_011517106.1
XM_017014849.1 Intron XP_016870338.1
XM_017014850.1 Intron XP_016870339.1
XM_017014851.1 Intron XP_016870340.1

View Full Product Details