Product Details

SNP ID
rs3923304
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:44809169 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTCCTTCTTTGGACTTTGGCCCC[C/T]GGCAGAAGAAAATGATTCTCAGCCC
Phenotype
MIM: 602913 MIM: 611713
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTBD19 PubMed Links

Gene Details

Gene
BTBD19
Gene Name
BTB domain containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136537.1 Intron NP_001130009.1
XM_006710386.3 Intron XP_006710449.1
XM_011540803.2 Intron XP_011539105.1
XM_011540805.2 Intron XP_011539107.1
XM_017000447.1 Intron XP_016855936.1
XM_017000448.1 Intron XP_016855937.1
XM_017000449.1 Intron XP_016855938.1
Gene
PLK3
Gene Name
polo like kinase 3
There are no transcripts associated with this gene.

Gene
TCTEX1D4
Gene Name
Tctex1 domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013632.3 Intron NP_001013654.1

View Full Product Details