Product Details

SNP ID
rs12726651
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:2642464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTGGGGCTGCTGGACGGAGGG[T/C]ACAGCCTGTGCTGTGGGCTGCTGCT
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
MMEL1 PubMed Links

Gene Details

Gene
MMEL1
Gene Name
membrane metallo-endopeptidase-like 1
There are no transcripts associated with this gene.

Gene
TTC34
Gene Name
tetratricopeptide repeat domain 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242672.1 Intron NP_001229601.1
XM_016999990.1 Intron XP_016855479.1

View Full Product Details