Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mrp-5
Entrez Gene ID
181587
Gene Name
Multidrug resistance-associated protein 5
Gene Aliases
CELE_F14F4.3
Chromosome Location
Chr.X: 14938502 - 14946319 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078078.3 NP_510479.1 18 - 19 3883 65

View Full Product Details