Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ekl-4
Entrez Gene ID
173310
Gene Name
protein-coding
Gene Aliases
CELE_Y105E8A.17
Chromosome Location
Chr.I: 14465168 - 14476894 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129058.4 NP_001122530.1 2 - 3 347 103
NM_170949.7 NP_740945.1 3 - 4 487 103

View Full Product Details