Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-45
Entrez Gene ID
177436
Gene Name
protein-coding
Gene Aliases
CELE_Y73B6A.5
Chromosome Location
Chr.IV: 6742537 - 6748572 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306912.2 NP_001293841.1 3 - 4 526 68
NM_001306913.3 NP_001293842.1 4 - 5 592 68
NM_001306914.2 NP_001293843.1 3 - 4 490 68
NM_001380288.1 NP_001368430.1 2 - 3 291 68
NM_001383109.1 4 - 5 549 68
NM_171367.8 NP_741430.3 3 - 4 423 68
GenBank mRNA AY455928.1 - 3 - 4 485 68
AY493413.1 - 3 - 4 477 68
AY493414.1 - 2 - 3 306 68

View Full Product Details