Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-45
Entrez Gene ID
177436
Gene Name
protein-coding
Gene Aliases
CELE_Y73B6A.5
Chromosome Location
Chr.IV: 6742537 - 6748572 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306912.2 NP_001293841.1 7 - 8 1422 79
NM_001306913.3 NP_001293842.1 8 - 9 1488 79
NM_001306914.2 NP_001293843.1 7 - 8 1386 79
NM_001380288.1 NP_001368430.1 6 - 7 1187 79
NM_001383109.1 8 - 9 1445 79
NM_171367.8 NP_741430.3 7 - 8 1319 79
GenBank mRNA AY455928.1 - 7 - 8 1381 79
AY493413.1 - 7 - 8 1373 79
AY493414.1 - 6 - 7 1202 79

View Full Product Details