Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
csn-2
Entrez Gene ID
172278
Gene Name
COP9 signalosome complex subunit 2
Gene Aliases
CELE_B0025.2
Chromosome Location
Chr.I: 6022667 - 6026877 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059339.9 NP_491740.1 3 - 4 729 105

View Full Product Details