Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rab-10
Entrez Gene ID
266836
Gene Name
Ras-related protein Rab-10
Gene Aliases
CELE_T23H2.5
Chromosome Location
Chr.I: 6457061 - 6458479 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059456.6 NP_491857.1 3 - 4 527 73

View Full Product Details