Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smg-1
Entrez Gene ID
172418
Gene Name
protein-coding
Gene Aliases
CELE_C48B6.6
Chromosome Location
Chr.I: 6900943 - 6913579 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025882.4 NP_001021053.1 31 - 32 5211 104
NM_001374908.2 NP_001361976.1 31 - 31 5189 104
GenBank mRNA AF149821.1 - 31 - 32 5211 104

View Full Product Details