Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smg-1
Entrez Gene ID
172418
Gene Name
protein-coding
Gene Aliases
CELE_C48B6.6
Chromosome Location
Chr.I: 6900943 - 6913579 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025882.4 NP_001021053.1 4 - 5 584 146
NM_001374908.2 NP_001361976.1 4 - 4 556 146
GenBank mRNA AF149821.1 - 4 - 5 584 146

View Full Product Details