Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
goa-1
Entrez Gene ID
172505
Gene Name
Guanine nucleotide-binding protein G(o) subunit alpha
Gene Aliases
CELE_C26C6.2
Chromosome Location
Chr.I: 7522116 - 7526716 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059707.6 NP_492108.1 2 - 3 395 71
GenBank mRNA AY008140.1 - 2 - 3 301 71

View Full Product Details