Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tsp-15
Entrez Gene ID
192065
Gene Name
Tetraspanin-15
Gene Aliases
CELE_F53B6.1
Chromosome Location
Chr.I: 8928748 - 8931089 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060003.6 NP_492404.1 2 - 3 286 152

View Full Product Details