Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lim-9
Entrez Gene ID
3565712
Gene Name
protein-coding
Gene Aliases
CELE_F25H5.1
Chromosome Location
Chr.I: 9181810 - 9220728 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026238.5 NP_001021409.2 1 - 2 187 83

View Full Product Details