Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lim-9
Entrez Gene ID
3565712
Gene Name
protein-coding
Gene Aliases
CELE_F25H5.1
Chromosome Location
Chr.I: 9181810 - 9220728 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026237.7 NP_001021408.2 1 - 2 167 74
NM_001026238.5 NP_001021409.2 5 - 6 840 74
NM_001030057.4 NP_001025228.2 7 - 8 1420 74
NM_001313465.3 NP_001300394.1 12 - 12 1586 74
NM_001313466.5 NP_001300395.1 13 - 13 1676 74
NM_001377616.3 NP_001364651.1 10 - 11 1265 74
NM_001377617.3 NP_001364650.1 7 - 8 989 74
GenBank mRNA EF443133.1 - 10 - 11 1265 74

View Full Product Details