Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lim-9
Entrez Gene ID
3565712
Gene Name
protein-coding
Gene Aliases
CELE_F25H5.1
Chromosome Location
Chr.I: 9181810 - 9220728 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313465.3 NP_001300394.1 6 - 6 742 69
NM_001313466.5 NP_001300395.1 7 - 7 832 69
NM_001377616.3 NP_001364651.1 4 - 5 421 69
NM_001377617.3 NP_001364650.1 1 - 2 145 69
GenBank mRNA EF443133.1 - 4 - 5 421 69

View Full Product Details