Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
irx-1
Entrez Gene ID
172789
Gene Name
Putative iroquois-class homeodomain protein irx-1
Gene Aliases
CELE_C36F7.1
Chromosome Location
Chr.I: 9536465 - 9540972 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060132.6 NP_492533.2 2 - 3 566 64

View Full Product Details