Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sur-6
Entrez Gene ID
172826
Gene Name
Serine/threonine-protein phosphatase 2A regulatory subunit sur-6
Gene Aliases
CELE_F26E4.1
Chromosome Location
Chr.I: 9754122 - 9757907 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060190.7 NP_492591.1 2 - 3 408 65
GenBank mRNA AF174643.1 - 2 - 3 406 65

View Full Product Details