Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mut-16
Entrez Gene ID
172873
Gene Name
protein-coding
Gene Aliases
CELE_B0379.3
Chromosome Location
Chr.I: 10080159 - 10085777 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060259.6 NP_492660.3 4 - 5 673 148
NM_060260.9 NP_492661.3 4 - 5 661 148

View Full Product Details