Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
che-13
Entrez Gene ID
186607
Gene Name
protein-coding
Gene Aliases
CELE_F59C6.7
Chromosome Location
Chr.I: 10506998 - 10515586 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001264468.3 NP_001251397.1 3 - 4 388 99
GenBank mRNA AF520567.1 - 3 - 4 308 99

View Full Product Details