Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tub-2
Entrez Gene ID
171758
Gene Name
protein-coding
Gene Aliases
CELE_Y71G12A.3
Chromosome Location
Chr.I: 1874008 - 1884196 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306423.3 NP_001293352.1 1 - 2 545 94
NM_058512.5 NP_490913.2 4 - 5 2334 94

View Full Product Details