Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zwl-1
Entrez Gene ID
171798
Gene Name
protein-coding
Gene Aliases
CELE_Y39G10AR.2
Chromosome Location
Chr.I: 2402230 - 2409999 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001262221.3 NP_001249150.1 5 - 6 1065 97
NM_001306389.3 NP_001293318.1 1 - 2 99 97

View Full Product Details