Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-11
Entrez Gene ID
171954
Gene Name
Nuclear Pore complex Protein
Gene Aliases
CELE_F53F10.5
Chromosome Location
Chr.I: 3818074 - 3821508 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058831.7 NP_491232.1 3 - 4 1687 65

View Full Product Details