Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pash-1
Entrez Gene ID
172950
Gene Name
protein-coding
Gene Aliases
CELE_T22A3.5
Chromosome Location
Chr.I: 10598673 - 10606621 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306532.2 NP_001293461.1 5 - 6 733 90

View Full Product Details