Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dmsr-1
Entrez Gene ID
186437
Gene Name
protein-coding
Gene Aliases
CELE_F57B7.1
Chromosome Location
Chr.V: 11438764 - 11443450 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171541.6 NP_741629.1 4 - 5 1169 72
NM_171542.7 NP_741630.1 4 - 5 1175 72

View Full Product Details