Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bed-1
Entrez Gene ID
180257
Gene Name
protein-coding
Gene Aliases
CELE_Y39B6A.12
Chromosome Location
Chr.V: 19119527 - 19124922 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029056.5 NP_001024227.1 3 - 4 465 134
NM_001029057.2 NP_001024228.1 3 - 4 458 134

View Full Product Details