Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lntl-1
Entrez Gene ID
183098
Gene Name
protein-coding
Gene Aliases
CELE_C31H1.6
Chromosome Location
Chr.IV: 5800336 - 5807093 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380241.1 NP_001367493.1 6 - 7 749 67
NM_001380242.1 NP_001368410.1 3 - 4 287 67
NM_068465.6 NP_500866.2 7 - 8 927 67
NR_131524.1 5 - 6 1068 67

View Full Product Details