Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gmd-2
Entrez Gene ID
186416
Gene Name
GDP-mannose 4,6 dehydratase 2
Gene Aliases
CELE_F56H6.5
Chromosome Location
Chr.I: 12292202 - 12293936 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060705.3 NP_493106.1 2 - 3 131 62

View Full Product Details