Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
amx-2
Entrez Gene ID
173146
Gene Name
Amino_oxidase domain-containing protein
Gene Aliases
CELE_B0019.1
Chromosome Location
Chr.I: 12759581 - 12764960 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383739.1 2 - 3 319 70

View Full Product Details