Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
math-48
Entrez Gene ID
191261
Gene Name
MATH (meprin-associated Traf homology) domain containing
Gene Aliases
CELE_ZK250.6
Chromosome Location
Chr.II: 1945438 - 1946527 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_061735.5 NP_494136.2 1 - 2 126 123

View Full Product Details