Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-10
Entrez Gene ID
173680
Gene Name
BTB and MATH domain containing
Gene Aliases
CELE_Y49F6C.4
Chromosome Location
Chr.II: 3375079 - 3376721 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062122.5 NP_494523.1 1 - 2 171 84

View Full Product Details