Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lact-5
Entrez Gene ID
183166
Gene Name
protein-coding
Gene Aliases
CELE_C33F10.7
Chromosome Location
Chr.II: 4818572 - 4821739 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026844.2 NP_001022015.1 5 - 6 849 136
NM_001026845.4 NP_001022016.1 4 - 5 602 136

View Full Product Details