Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ggr-3
Entrez Gene ID
191638
Gene Name
GABA/Glycine Receptor family (see gbr)
Gene Aliases
CELE_F09C12.1
Chromosome Location
Chr.II: 5130381 - 5133472 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062550.5 NP_494951.3 3 - 4 295 104

View Full Product Details