Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acdh-8
Entrez Gene ID
173979
Gene Name
Acyl CoA DeHydrogenase
Gene Aliases
CELE_K05F1.3
Chromosome Location
Chr.II: 5794495 - 5796185 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062741.7 NP_495142.1 3 - 4 1135 84

View Full Product Details