Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpy-1
Entrez Gene ID
174105
Gene Name
43 kDa receptor-associated protein of the synapse homolog
Gene Aliases
CELE_C18H9.7
Chromosome Location
Chr.II: 6702213 - 6705657 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383869.1 4 - 5 547 105

View Full Product Details