Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acdh-12
Entrez Gene ID
174180
Gene Name
Acyl CoA DeHydrogenase
Gene Aliases
CELE_E04F6.5
Chromosome Location
Chr.II: 7192429 - 7195641 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026891.6 NP_001022062.1 8 - 9 1748 110
NR_151558.1 7 - 8 1403 110

View Full Product Details