The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322749.3 | NP_001309578.1 | 5 - 5 | 606 | 67 |
NM_001322750.3 | NP_001309579.1 | 2 - 3 | 188 | 67 |