Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sec-5
Entrez Gene ID
174459
Gene Name
Exocyst complex component 2
Gene Aliases
CELE_T23G7.4
Chromosome Location
Chr.II: 9173161 - 9177152 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063557.6 NP_495958.1 6 - 7 1394 70

View Full Product Details