Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- nuo-1
- Entrez Gene ID
-
174696
- Gene Name
- NADH dehydrogenase [ubiquinone] flavoprotein 1, mitochondrial
- Gene Aliases
-
CELE_C09H10.3
- Chromosome Location
-
Chr.II: 11097837 - 11100146 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063975.5 |
NP_496376.1 |
2 - 3 |
219 |
95 |
- Gene Symbol
- C09H10.4
- Entrez Gene ID
-
13187006
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_C09H10.4
- Chromosome Location
-
Chr.II: 11097850 - 11100497 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383946.1 |
|
2 - 3 |
206 |
95 |
View Full Product Details