Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-5
Entrez Gene ID
174779
Gene Name
Nuclear pore complex protein 5
Gene Aliases
CELE_F07A11.3
Chromosome Location
Chr.II: 11596767 - 11599809 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064080.6 NP_496481.1 4 - 5 1461 119

View Full Product Details