Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-37
Entrez Gene ID
184512
Gene Name
Serpentine receptor class epsilon-37
Gene Aliases
CELE_F15A4.3
Chromosome Location
Chr.II: 12465857 - 12467533 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064254.3 NP_496655.2 2 - 3 784 146

View Full Product Details