Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ztf-18
Entrez Gene ID
175250
Gene Name
C2H2-type domain-containing protein
Gene Aliases
CELE_T24C4.7
Chromosome Location
Chr.III: 891326 - 895712 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381718.1 NP_001367780.1 7 - 8 2044 109

View Full Product Details