Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rep-1
Entrez Gene ID
175311
Gene Name
Rab GDP dissociation inhibitor
Gene Aliases
CELE_Y67D2.1
Chromosome Location
Chr.III: 1570306 - 1576649 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027757.5 NP_001022928.1 2 - 3 205 135

View Full Product Details