Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-62
Entrez Gene ID
183322
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_C39B5.3
Chromosome Location
Chr.III: 2227707 - 2229268 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065115.4 NP_497516.2 2 - 3 686 115

View Full Product Details