Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ben-1
Entrez Gene ID
175461
Gene Name
Tubulin beta chain
Gene Aliases
CELE_C54C6.2
Chromosome Location
Chr.III: 3537642 - 3541639 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065327.4 NP_497728.2 1 - 2 213 81

View Full Product Details